Learn More
CPC Scientific H-Cys-Lys-Ala-Thr-Gln-Ala-Ser-Gln-Glu-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST013A
Encompass
This peptide is the last 9 C-terminal residues of histone H2AX and a C-terminal cysteine residue for conjugation. Histone H2AX is a member of the histone H2A family that differs from other H2A histones by the presence of this C-terminal sequence.ONE-LETTER SEQUENCE: CKATQASQEYMOLECULAR FORMULA: C46H73N13O18MOLECULAR WEIGHT:1128.22STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Rogakou, E. et al. J Biol Chem 275, 9390 (2000)
Catalog No. 50-210-9443