Learn More
Enzo Life Sciences Histone H2B (human), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKPRCC001F
Encompass_Preferred
Purity: ≥95% (SDS-PAGE; Western blot). MW: ~15kDa. Formulation: Liquid. In 1x Dulbecco's PBS. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Kinase assay.
Catalog No. 50-200-7325