Learn More
ECM Bioscience Histone H2B (Ser-36), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience HP4331
Encompass_Preferred
The antibody detects a 15 kDa* protein corresponding to the molecular mass of Histone H2B on SDS-PAGE immunoblots of human Jurkat cells treated with Calyculin A, and this reactivity is reduced after lambda phosphatase treatment. The immunogen was Phospho-Histone H2B (Ser-36) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding Ser-36 in human histone H2B. This sequence is well conserved in most isoforms of rat and mouse histone H2B, and has low homology to other histone family members. Accession #P33778. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = HIST1H2BB, H2BFF, Histone H2B type1B, H2B/f
Catalog No. 50-172-0795