Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Ala-Thr-Lys-Ala-Ala-Arg-Lys-Ser-Ala-Pro-Ala-Thr-Gly-Gly-Val-OH (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST039B
Encompass
This peptide is the fragment of Histone H3 (1-35).ONE-LETTER SEQUENCE: ARTKQTARKSTGGKAPRKQLATKAARKSAPATGGVMOLECULAR FORMULA: C149H269N55O45MOLECULAR WEIGHT:3551.09STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9477