Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Ala-Thr-Lys-Ala-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST036A
Encompass
This peptide is the N-terminus of histone H3 (1-25), amidated. ONE-LETTER SEQUENCE: ARTKQTARKSTGGKAPRKQLATKAA-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:2625.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Kobza, K. et al. FEBS J. 272, 4249 (2005)
Catalog No. 50-210-9471