Learn More
Cayman Chemical Histone H3 (Citrullinated R2 + R8 + R17) Monoclonal Antibody, 100 µg
SDP
Supplier:  Cayman Chemical 17939100
Encompass_Preferred
Antigen: histone 3 peptide with citrullinations at R2, R8, and R17 • Isotype: IgG1 • Host: rabbit • Species Reactivity: (+) human • Application(s): ELISA and WB
Catalog No. NC1103176