Learn More
Enzo Life Sciences Histone H3 dimethyl lysine-4 peptide (0.5mg)
SDP
Catalog No. 502011316
Encompass_Preferred
Residues 1-21 from the N-terminal tail of human histone H3, dimethylated on the side-chain (ε-amino function) of Lys4. Useful as a substrate for histone lysine demethylases, for example LSD1. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1316 Supplier Enzo Life Sciences Supplier No. BMLP2560500
Residues 1-21 from the N-terminal tail of human histone H3, dimethylated on the side-chain (ε-amino function) of Lys4. Useful as a substrate for histone lysine demethylases, for example LSD1. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.