Learn More
CPC Scientific H-Ala-Thr-Lys-Ala-Ala-Arg-Lys-Ser-Ala-Pro-Ser-Thr-Gly-Gly-Val-Lys-Lys-Pro-His-Arg-Tyr-Arg-Pro-Gly-Gly-Lys(Biotin)-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST050B
Encompass
This peptide is the fragment of Histone H3 (21-44) with an additional C-terminal Gly followed by a biotinylated Lys, amidated.ONE-LETTER SEQUENCE: ATKAARKSAPSTGGVKKPHRYRPG-GK(Biotin)-NH2MOLECULAR FORMULA: C127H215N45O33S1MOLECULAR WEIGHT:2932.43STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9492