Learn More
Enzo Life Sciences Histone H3 (human) (5-23) amide (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP2710500
Encompass_Preferred
Comprising residues 5-23 of the human histone H3 N-terminal tail, this peptide is centered on Lys14, preferred acetylation site for the GCN5/PCAF family of histone acetyltransferases (HATs). May be used as a substrate for assay of PCAF and hGCN5 HATs. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1327