Learn More
Cayman Chemical Histone H3 (human recombinant), 1 mg
SDP
Supplier:  Cayman Chemical 102631
Encompass_Preferred
Source: Recombinant protein expressed in E. coli • Mr: 15.5 kDa • Histone H3 is one of the core nucleosomal histones. It undergoes many modifications which include acetylation, methylation, and phosphorylation that are important for regulation of gene transcription.
Catalog No. NC0791699