Learn More
Medchemexpress LLC Histone H3 Human 9 250ug
SDP
Supplier:  Medchemexpress LLC HYP72335250UG
Encompass_Preferred
Histone H3 proteins are critical in nucleosomes which compact DNA into chromatin and regulate DNA accessibility Histone H3 Protein Human (98a a His) is the recombinant human-derived Histone H3 protein expressed by E coli with C-6 His labeled tag
Catalog No. 50-001-82687