Learn More
Medchemexpress LLC Histone H3 Human H 5ug
SDP
Catalog No. 5000181850
Encompass_Preferred
Histone H3 proteins are critical in nucleosomes which compact DNA into chromatin and regulate DNA accessibility Histone H3 Protein Human (His) is the recombinant human-derived Histone H3 protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-81850 Supplier Medchemexpress LLC Supplier No. HYP727985UG
Histone H3 proteins are critical in nucleosomes which compact DNA into chromatin and regulate DNA accessibility Histone H3 Protein Human (His) is the recombinant human-derived Histone H3 protein expressed by E coli with N-His labeled tag