Learn More
LICOR Histone H3 Mouse Monoclonal Antibody for Normalization (100 uL)
Supplier:  LICOR 92642218
Encompass_Preferred
The Histone H3 primary antibody can be used as an internal loading control for normalization and is particularly effective when detecting target proteins in nuclear extracts. The expression of Histone H3, or any housekeeping protein (HKP), should be validated to ensure that its expression does not change under experimental conditions. Once validated, Histone H3 primary antibodies can be used for the detection of Histone H3 when performing multiplex Western blot detection. Detect Histone H3 Mouse Monoclonal Antibody with IRDye Goat anti-Mouse or IRDye Donkey anti-Mouse secondary antibodies.
Catalog No. NC2209188