Learn More
Active Motif Histone H3 trimethyl Lys9 ELISA
SDP
Supplier:  Active Motif 53109
Encompass
Histone Modification ELISAs provide a simple, sensitive method for detecting changes in histone methylation levels from purified core histones or histones isolated by acid extraction. For added convenience and a more quantitative interpretation of results, the Histone Modification ELISAs also include a site- and degree-specific methylated recombinant histone protein for use as a reference standard curve.
Catalog No. NC1852623