Learn More
Cayman Chemical Histone H3 Trimethyl Lys9 Peptide, 1 mg
SDP
Catalog No. 502887622
Encompass_Preferred
Peptide contains a trimethylated lysine residue at position nine which is a substrate for several of the JMJC domain-containing class of histone demethylases; JMJD2A and JMJD2D exhibit higher affinity with H3K9me3 compared to H3K9me2
Catalog No. 50-288-7622 Supplier Cayman Chemical Supplier No. 105301 MG
Peptide contains a trimethylated lysine residue at position nine which is a substrate for several of the JMJC domain-containing class of histone demethylases; JMJD2A and JMJD2D exhibit higher affinity with H3K9me3 compared to H3K9me2