Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Trp-Cys-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109457
Encompass
This is an amidated sequence of Histone H3 (1-18) with two additional residues (WC) on its C-terminus. ONE-LETTER SEQUENCE: ARTKQTARKSTGGKAPRKWC-NH2MOLECULAR FORMULA: C94H164N36O25S1MOLECULAR WEIGHT:2230.61STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9457 Supplier CPC Scientific Supplier No. HIST024A
This is an amidated sequence of Histone H3 (1-18) with two additional residues (WC) on its C-terminus. ONE-LETTER SEQUENCE: ARTKQTARKSTGGKAPRKWC-NH2MOLECULAR FORMULA: C94H164N36O25S1MOLECULAR WEIGHT:2230.61STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous