Learn More
Medchemexpress LLC Histone H3 Xenopus 10ug
SDP
Catalog No. 5000182329
Encompass_Preferred
Histone H3 Protein constitutes a core element in the nucleosome an octamer comprising H2A H2B H3 and H4 The assembly forms a histone octamer with one H3-H4 heterotetramer and two H2A-H2B heterodimers serving as a molecular spool that wraps around 147 base pairs of DNA This compact organization contributes to chromatin structure and Histone H3 as part of this assembly plays a crucial role in chromatin structure and gene regulation Histone H3 Protein Xenopus laevis (135a a) is the recombinant Xenopus laevis-derived Histone H3 protein expressed by E coli with tag free
Catalog No. 50-001-82329 Supplier Medchemexpress LLC Supplier No. HYP7233310UG
Histone H3 Protein constitutes a core element in the nucleosome an octamer comprising H2A H2B H3 and H4 The assembly forms a histone octamer with one H3-H4 heterotetramer and two H2A-H2B heterodimers serving as a molecular spool that wraps around 147 base pairs of DNA This compact organization contributes to chromatin structure and Histone H3 as part of this assembly plays a crucial role in chromatin structure and gene regulation Histone H3 Protein Xenopus laevis (135a a) is the recombinant Xenopus laevis-derived Histone H3 protein expressed by E coli with tag free