Learn More
Epigentek Inc Histone H3K27me3 (H3K27 Trimethyl) Polyclonal Antibody (25 ul)
SDP
Supplier:  Epigentek Inc A4039025
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Synthetic Peptide of Human TriMethyl-Histone H3-K27. Tested Applications: WB, IHC, IF, IP, ChIP, ChIPseq. Reactivity: Broad Range, Mouse, Rat, Human. Alternative Names: HIST3H3, H3/g.
Catalog No. 50-109-5518