Learn More
Active Motif Histone H3K27me1 Peptide - biotinylated
SDP
Supplier:  Active Motif 81050
Encompass
Histone H3K27me1 Peptide for the study of proteins that interact with histones and that read, write or erase histone modifications.
Catalog No. NC1851344