Learn More
Medchemexpress LLC Histone H4 Human/Xe 5ug
SDP
Catalog No. 5000181953
Encompass_Preferred
Histone H4 proteins are core components of nucleosomes which compact DNA into chromatin and regulate DNA accessibility during cellular processes Histones including H4 play critical roles in transcriptional regulation DNA repair replication and chromosome stability Histone H4 Protein Human/Xenopus laevis is the recombinant Xenopus laevis-derived Histone H4 protein expressed by E coli with tag free
Catalog No. 50-001-81953 Supplier Medchemexpress LLC Supplier No. HYP723365UG
Histone H4 proteins are core components of nucleosomes which compact DNA into chromatin and regulate DNA accessibility during cellular processes Histones including H4 play critical roles in transcriptional regulation DNA repair replication and chromosome stability Histone H4 Protein Human/Xenopus laevis is the recombinant Xenopus laevis-derived Histone H4 protein expressed by E coli with tag free