Learn More
Proteintech Group Inc Histone-Label Atto488
SDP
Supplier:  Proteintech Group Inc TBA488100
Encompass
Histone-Label* is a fluorescent chromatin probe for direct staining of chromosomes and nuclei. It consists of an anti-histone VHH conjugated to the fluorescent dye ATTO488. For IF, microscopy of histones, chromosomes, and nuclei.
Catalog No. 50-225-8870