Learn More
Sigma Aldrich Fine Chemicals Biosciences Histone Octamer full lengt
Catalog No. 502907052
Encompass
Human recombinant histone octamer consisting of 2 molecules each of histones H2A (GenBank Accession No. NM 033445) amino acids 2-130(end) with a N-terminal His-tag H2B (GenBank Accession No. NM 003528) amino acids 2-126(end) with a N-terminal His-tag H3 (GenBank Accession No. NM 003532) amino acids 2-137(end) with a N-terminal His-tag and H4 (GenBank Accession No. NM 003548) amino acids 2-103(end) with a N-terminal His-tag expressed in an E. coli expression system.
Catalog No. 50-290-7052 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP0408100UG
Human recombinant histone octamer consisting of 2 molecules each of histones H2A (GenBank Accession No. NM 033445) amino acids 2-130(end) with a N-terminal His-tag H2B (GenBank Accession No. NM 003528) amino acids 2-126(end) with a N-terminal His-tag H3 (GenBank Accession No. NM 003532) amino acids 2-137(end) with a N-terminal His-tag and H4 (GenBank Accession No. NM 003548) amino acids 2-103(end) with a N-terminal His-tag expressed in an E. coli expression system.