Learn More
EPICYPHER INC HISTONE OCTAMER, RECOMB HUM,50
SDP
Supplier:  EPICYPHER INC 160001
Encompass
Histone Octamer, Recombinant Human, 50 μgHistone Octamer, Recombinant Human$210.00SKU: 16-0001Pack Size: 50 μgSelect Pack Size * 50 µg1Add to Cart Share: Histone Octamer, Recombinant Human Description: Human histone octamers (two each of histones H2A, H2B, H3 and H4) made from recombinant histones expressed in E. coli (accession numbers: H2A-P0C0S8; H2B-P62807; H3-P68431; H4-P62805). The histone octamer is the protein component of the nucleosome, the basic subunit of chromatin. A nucleosome consists of a histone octamer wrapped with 147 base pairs of DNA.
Catalog No. NC1835923
 Restricted Item