Learn More
Active Motif Histone Purification Mini Kit
SDP
Catalog No. NC9774719
Encompass
Active Motif's Histone Purification Mini Kit enables you to purify core histones (H2A, H2B, H3 and H4) as one total population while preserving their post-translational modifications such as acetylation, methylation, and phosphorylation states. This purification method is an improvement over acid precipitation methods and utilizes a convenient spin column and proprietary buffer system to purify the core histones from cells and tissue samples. Histones isolated by this method are suitable substrates for downstream assays and in vitro chromatin assembly.
Catalog No. NC9774719 Supplier Active Motif Supplier No. 40026
Active Motif's Histone Purification Mini Kit enables you to purify core histones (H2A, H2B, H3 and H4) as one total population while preserving their post-translational modifications such as acetylation, methylation, and phosphorylation states. This purification method is an improvement over acid precipitation methods and utilizes a convenient spin column and proprietary buffer system to purify the core histones from cells and tissue samples. Histones isolated by this method are suitable substrates for downstream assays and in vitro chromatin assembly.