Learn More
Sigma Aldrich Fine Chemicals Biosciences Histopaque-1119, 6 x 100 mL
Catalog No. NC1924361
Encompass
A solution containing polysucrose and sodium diatrizoate, adjusted to a density of 1.119 g/mL. Combined with Histopaque-1077, it permits the separation of mononuclear cells and granulocytes.
Catalog No. NC1924361 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 111916X100ML
A solution containing polysucrose and sodium diatrizoate, adjusted to a density of 1.119 g/mL. Combined with Histopaque-1077, it permits the separation of mononuclear cells and granulocytes.