Learn More
Alkali Scientific Histoplast Paraffin Wax Ultra-
SDP
Catalog No. 503028266
Encompass_Preferred
Alkali Scientificpremium paraffin wax blended with purified plastic polymers is specifically designed for general use and all histology applications With a melting point of 56C to 58C it ensures optimal performance for detailed sectioning tasks The DMSO-free formulation enhances safety and reliability minimizing compression and cell disruption to produce high-quality slides This wax supports ultra-thin sectioning down to 2 m with exceptional precision Its convenient pearl-shaped granules simplify handling and melting making it a versatile and efficient choice for a variety of laboratory protocols High-quality paraffin wax with purified plastic polymers Suitable for general use and all histology applications DMSO-free for safer use and improved performance Ensures ultra-smooth cutting with minimal compression and cell disruption Supports precise sectioning as thin as 2 m for detailed results Pearl-shaped granules for effortless handling and efficient melting Melting point of 56C
Catalog No. 50-302-8266 Supplier Alkali Scientific Supplier No. PFW01CS
Alkali Scientificpremium paraffin wax blended with purified plastic polymers is specifically designed for general use and all histology applications With a melting point of 56C to 58C it ensures optimal performance for detailed sectioning tasks The DMSO-free formulation enhances safety and reliability minimizing compression and cell disruption to produce high-quality slides This wax supports ultra-thin sectioning down to 2 m with exceptional precision Its convenient pearl-shaped granules simplify handling and melting making it a versatile and efficient choice for a variety of laboratory protocols High-quality paraffin wax with purified plastic polymers Suitable for general use and all histology applications DMSO-free for safer use and improved performance Ensures ultra-smooth cutting with minimal compression and cell disruption Supports precise sectioning as thin as 2 m for detailed results Pearl-shaped granules for effortless handling and efficient melting Melting point of 56C