Learn More
Epredia™ Histoplast Paraffin
Catalog No. 8330 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Paraffin IM
Paraffin Kit LP
Paraffin Kit PE
Paraffin LP
Paraffin PE
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
83-30 Paraffin PE
8330K Paraffin Kit PE
83-31 Paraffin IM
83-32 Paraffin LP
8332K Paraffin Kit LP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 83-30 Supplier Epredia™ Supplier No. 8330
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.
  • Made of paraffin and plastic polymers
  • Delivers excellent processing and yields thin, wrinkle-free sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).
Specifications
Melting Point 56°C
Product Type Paraffin PE
Quantity 8 x 1 Kg
WARNING: Cancer - www.P65Warnings.ca.gov