Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HistoPrep™ 2-Propanol, Fisherbrand™
SDP

Catalog No. 11002211 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-211 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-211
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

99% absolute isopropyl alcohol.

  • A dehydrating agent during tissue processing, specifically for dehydrating tissue samples before embedding them in paraffin for sectioning
  • Ideal for small tissue, biopsy and microwave processing.

Chemical Identifiers

CAS 67-63-0

Specifications

Physical Form Aqueous Solution
Color Clear
Quantity 1 gal.
Percent Purity 100%
Grade Reagent

Made in the USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.