Learn More
Electron Microscopy Sciences HISTOPRO RAPID TISSUE PROCESS
SDP
Catalog No. NC3401315
Encompass
  • Rapid processing of small biopsies in under 75 minutes.
  • High throughput capacity handling over 2,500 cassettes in an 8-hour shift.
  • Artifact-free results using a method that avoids microwave artifacts.
  • Continuous load system allows uninterrupted addition of new samples.
  • Compact design suitable for various laboratory environments.
  • Integrated fume control with an activated charcoal filter.
  • User-friendly interface with an intuitive LCD display.
  • Cost-effective operation using a regular power outlet.
  • Manufactured in the USA for quality and reliability.
Catalog No. NC3401315 Supplier Electron Microscopy Sciences Supplier No. 6258110
Add to Cart
Add to Cart
Request a Quote
  • Rapid processing of small biopsies in under 75 minutes.
  • High throughput capacity handling over 2,500 cassettes in an 8-hour shift.
  • Artifact-free results using a method that avoids microwave artifacts.
  • Continuous load system allows uninterrupted addition of new samples.
  • Compact design suitable for various laboratory environments.
  • Integrated fume control with an activated charcoal filter.
  • User-friendly interface with an intuitive LCD display.
  • Cost-effective operation using a regular power outlet.
  • Manufactured in the USA for quality and reliability.