Learn More
StatLab Histosette II Embedding Cassettes
SDP
Catalog No. 21033424
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-033-424 Blue
21-033-419 Orange
21-033-420 White
21-033-421 Pink
21-033-422 Green
21-033-423 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 21-033-424 Supplier StatLab Supplier No. M4856SL
Add to Cart
Add to Cart
Unique front-hinged routine cassette design with a rear tab that allows for unobstructed access to the front labeling surface
  • Sleeved, CS750
  • Lids Separate
Specifications
Color Blue
Product Type Embedding Cassette