Learn More
Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes with Combination Lid and Base
Catalog No. 15182701B Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-182-701B Pink
15-182-701A White
15-182-701C Green
15-182-701D Yellow
15-182-701E Blue
15-182-701F Peach
15-182-701G Tan
15-182-701H Gray
15-182-701J Lilac
15-182-701K Orange
15-182-701L Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 15-182-701B
Add to Cart
Add to Cart
This item is not returnable. View return policy
Histosette™ II Tissue Processing/Embedding Cassettes with Combination Lid and Base open and close as often as necessary - cassettes will always relock securely.
  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Pore Style Slots
Writing Face Angle 45°
Closure Type Combination Lid
Color Pink
Material Plastic
Product Type Tissue Cassette