Learn More
Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes with Combination Lid and Base
Catalog No. 15182701G Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Catalog No. Color
15-182-701G Tan
15-182-701A White
15-182-701B Pink
15-182-701C Green
15-182-701D Yellow
15-182-701E Blue
15-182-701F Peach
15-182-701H Gray
15-182-701J Lilac
15-182-701K Orange
15-182-701L Aqua
11 options
This item is not returnable. View return policy
Catalog No. 15-182-701G
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Histosette™ II Tissue Processing/Embedding Cassettes with Combination Lid and Base open and close as often as necessary - cassettes will always relock securely.
  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Pore Style Slots
Writing Face Angle 45°
Closure Type Combination Lid
Color Tan
Material Plastic
Product Type Tissue Cassette