Learn More
National Diagnostics Histosol, 1 Gallon
SDP
Catalog No. NC1706825
Encompass
Histosol is the original high flash point (114°F TCC) histological clearing agent. It is intended to be used as a replacement for xylene where the hazards associated with aromatic hydrocarbon vapors are to be reduced. Museum-quality tissue slides can be prepared with Histosol without change in protocol or procedure. Histosol is manufactured from petrochemical products and is miscible in all proportions with ethanol, isopropanol, and t-butanol. It is also miscible with all paraffin-based tissue embedding media and all permanent mounting materials.
This item is not returnable. View return policy
Catalog No. NC1706825 Supplier National Diagnostics Supplier No. HS100
Histosol is the original high flash point (114°F TCC) histological clearing agent. It is intended to be used as a replacement for xylene where the hazards associated with aromatic hydrocarbon vapors are to be reduced. Museum-quality tissue slides can be prepared with Histosol without change in protocol or procedure. Histosol is manufactured from petrochemical products and is miscible in all proportions with ethanol, isopropanol, and t-butanol. It is also miscible with all paraffin-based tissue embedding media and all permanent mounting materials.