Learn More
Electron Microscopy Sciences Histosol™ Histosol with Xylene Substitute 4 GL
SDP
Catalog No. 501901210
Encompass_Preferred
Histosol is the original high flash point (114 degree F TCC) histological clearing agent. It is intended to be used as a replacement for xylene where the hazards associated with aromatic hydrocarbon vapors are to be reduced. Museum-quality tissue slides can be prepared with Hitosol without change in protocol or procedure. Histosol is manufactured from petrochemical products and is miscible in all proportions with ethanol, isopropanol, and t-butanol. It is also miscible with all paraffin-based tissue embedding media and all permanent mounting materials.
Catalog No. 50-190-1210 Supplier Electron Microscopy Sciences Supplier No. 2340504
Histosol is the original high flash point (114 degree F TCC) histological clearing agent. It is intended to be used as a replacement for xylene where the hazards associated with aromatic hydrocarbon vapors are to be reduced. Museum-quality tissue slides can be prepared with Hitosol without change in protocol or procedure. Histosol is manufactured from petrochemical products and is miscible in all proportions with ethanol, isopropanol, and t-butanol. It is also miscible with all paraffin-based tissue embedding media and all permanent mounting materials.