Learn More
Newcomer Supply HistoTec Marking Pen, 12/box for painted end of slides and cassettes
SDP
Supplier:  Newcomer Supply 5725
Encompass
Fine-tip (diameter approximately 0.9mm), black ink pen that writes and dries quickly.  Works on microscope slides and plastic cassettes.  Withstands histo & cyto stains, xylene, alcohol and other histo chemistries.
Catalog No. NC9175748