Learn More
Nacalai USA Inc HISTOVT ONE
Catalog No. NC0499519
Encompass
HistoVT One (10x, pH 7.0), Room Temperature Storage, 500 ml. HistoVT One is an antigen retrieval solution for immunohistochemistry and in situ hybridization. This product can unmask antigenic sites without damage to antigen from formalin-fixed, frozen or paraffin-embedded tissue sections.
Catalog No. NC0499519 Supplier Nacalai USA Inc Supplier No. 0638005
HistoVT One (10x, pH 7.0), Room Temperature Storage, 500 ml. HistoVT One is an antigen retrieval solution for immunohistochemistry and in situ hybridization. This product can unmask antigenic sites without damage to antigen from formalin-fixed, frozen or paraffin-embedded tissue sections.