Learn More
Sigma Aldrich Fine Chemicals Biosciences HisTrapTM fast flow Cytiva
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17531901
Encompass
HisTrap(TM) ff is a ready-to-use column prepacked with precharged Ni Sepharose(R) 6 fast flow for preparative purification of histidine-tagged recombinant proteins by immobilized metal ion affinity chromatography (IMAC).Ni Sepharose(R) 6 fast flow consists of 90 mum beads of highly cross-linked agarose to which a chelating ligand has been immobilized. This chelating ligand is charged with Ni2 ions the first-choice metal ion for purifying most histidine-tagged proteins. The negligible leakage of Ni2 ions from the matrix ensures reliable capture of histidine-tagged proteins in repeated IMAC purifications.The high flow rates made possible by the Ni Sepharose(R) 6 fast flow matrix make HisTrap(TM) ff columns well suited for small scale-up. Capacity can be increased by connecting columns in series. HiTrap(R) IMAC ff is the product of choice when charging the medium with different metal ions for optimization of purification protocols.
Catalog No. 50-290-1610