Learn More
Sigma Aldrich Fine Chemicals Biosciences HisTrapTM High Performance
Catalog No. 502901597
Encompass
HisTrap(TM) HP columns are prepackedwith NiSepharose(R) High Performance and designed for simple high-reSOLUTIONon purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).HisTrap(TM) HP 1 mL and 5 mL columns are designed for simple one-step purification of histidine-tagged proteins. The columns are prepacked with Ni Sepharose(R) High Performance which has high binding capacity and low nickel ion leakage that ensures reliable capture of target protein in repeated IMAC purifications.HisTrap(TM) HP columns can also be used for the purification of tagged proteins containing shorter or longer polyhistidine tags such as (histidine)4 or (histidine)10. The shorter (histidine)4 will bind more weakly and the longer (histidine)10 will bind more strongly compared with (histidine)6. This difference in binding strength can be used during purification since (histidine)10 binds more strongly a higher concentration of imidazole can be added to the lysed ce
Catalog No. 50-290-1597 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17524801
HisTrap(TM) HP columns are prepackedwith NiSepharose(R) High Performance and designed for simple high-reSOLUTIONon purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).HisTrap(TM) HP 1 mL and 5 mL columns are designed for simple one-step purification of histidine-tagged proteins. The columns are prepacked with Ni Sepharose(R) High Performance which has high binding capacity and low nickel ion leakage that ensures reliable capture of target protein in repeated IMAC purifications.HisTrap(TM) HP columns can also be used for the purification of tagged proteins containing shorter or longer polyhistidine tags such as (histidine)4 or (histidine)10. The shorter (histidine)4 will bind more weakly and the longer (histidine)10 will bind more strongly compared with (histidine)6. This difference in binding strength can be used during purification since (histidine)10 binds more strongly a higher concentration of imidazole can be added to the lysed ce