Learn More
Medchemexpress LLC Histrelin acetate | 220810-26-4 | 99.9% | 1323.50 (free base) | 1 MG
SDP
Catalog No. 5000426680
Encompass_Preferred
Histrelin acetate is a GnRH analogue and a GnRH Receptor agonist. It increases serum luteinising hormone (LH), follicle stimulating hormone (FSH), and testosterone levels. It can be used in the research of prostate cancer and endometriosis.
  • Stimulates release of vasopressin (VP) from isolated rat hypothalamo-neurohypophysial explants
  • Stimulates oxytocin (OT) release from the rat hypothalamo-neurohypophysial system
  • Rescues circulating LH concentrations in Csfmop/Csfmop mice
  • Reduces the number of endometrial glands and atrophies the stroma in rabbits
Catalog No. 50-004-26680 Supplier Medchemexpress LLC Supplier No. HYP0056A1MG
Histrelin acetate is a GnRH analogue and a GnRH Receptor agonist. It increases serum luteinising hormone (LH), follicle stimulating hormone (FSH), and testosterone levels. It can be used in the research of prostate cancer and endometriosis.
  • Stimulates release of vasopressin (VP) from isolated rat hypothalamo-neurohypophysial explants
  • Stimulates oxytocin (OT) release from the rat hypothalamo-neurohypophysial system
  • Rescues circulating LH concentrations in Csfmop/Csfmop mice
  • Reduces the number of endometrial glands and atrophies the stroma in rabbits