Learn More
Ocean Nano Tech LLC HiSur Streptavidin Beads, 100 mL, Magnetic, Streptavidin, Hydrophilic, 1 μm, 10 mg/mL, Storage 2-8°C, Binding Capacity >50 μg biotinylated IgG / mg beads
SDP
Catalog No. NC2455742
Encompass
HiSur Streptavidin Beads, 100 mL, Magnetic, Streptavidin, Hydrophilic, 1 μm, 10 mg/mL, Storage 2-8°C, Binding Capacity >50 μg biotinylated IgG / mg beads
Catalog No. NC2455742 Supplier Ocean Nano Tech LLC Supplier No. HV1000100
HiSur Streptavidin Beads, 100 mL, Magnetic, Streptavidin, Hydrophilic, 1 μm, 10 mg/mL, Storage 2-8°C, Binding Capacity >50 μg biotinylated IgG / mg beads