Learn More
Analytical Sales & Services Hitachi Models M655/L6000/L620
Catalog No. 5000805609
Encompass
Hitachi Models M655/L6000/L6200/L6200A/L71000 Replacement Teflon Black Piston Seals OEM No. 655-1080
Qty/Pkg: 10PK
Catalog No. 50-008-05609 Supplier Analytical Sales & Services Supplier No. 900298
Add to Cart
Add to Cart
Hitachi Models M655/L6000/L6200/L6200A/L71000 Replacement Teflon Black Piston Seals OEM No. 655-1080
Qty/Pkg: 10PK