Learn More
Electron Microscopy Sciences Hitachi 3-Position Multi-Sample Holder
SDP
Catalog No. 501930543
Encompass_Preferred
A single tilt multi-grid holder. Will accommodate 3 grids with easy reproducible movement between each specimen. It is ideal for scanning through serial samples or for comparing against standards. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in a mahogany box.
Catalog No. 50-193-0543 Supplier Electron Microscopy Sciences Supplier No. EMS022
Add to Cart
Add to Cart
Request a Quote
A single tilt multi-grid holder. Will accommodate 3 grids with easy reproducible movement between each specimen. It is ideal for scanning through serial samples or for comparing against standards. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in a mahogany box.