Learn More
AdipoGen HITCI
SDP
Supplier:  AdipoGen CDXH0926G005
Encompass_Preferred
Chemical. CAS 19764-96-6. Formula C29H33IN2. MW 536.49. Indocarbocyanine dye used typically as a laser dye in the near infrared NIR. Spectral data Absorption lambdamax 740nm MeOH, EtOH and 751nm DMSO, Fluorescence lambdamax 778nm MeOH, EtOH and 792nm DMSO. This laser dye is for pulsed and continuous wave operation and tunable around 840-860nm.
Catalog No. 50-256-0683