Learn More
Ancare Corporation HITEMP POLY RAT CAGE BTM 10CS
SDP
Catalog No. NC1730107
Encompass
Hi-Temp Polycarbonate Cage Bottom, Rat• 19” x 10 ½” x 8”• Slightly tinted, autoclavable to 270ºF (131ºC)• 10 per case
Catalog No. NC1730107 Supplier Ancare Corporation Supplier No. R20HT
Hi-Temp Polycarbonate Cage Bottom, Rat• 19” x 10 ½” x 8”• Slightly tinted, autoclavable to 270ºF (131ºC)• 10 per case