Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Benzamidine fast f
Catalog No. 502904961
Encompass
HiTrap(R) Benzamidine ff (high sub) are prepacked with Benzamidine Sepharose(R) 4 fast fLow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.
Catalog No. 50-290-4961 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17514401
HiTrap(R) Benzamidine ff (high sub) are prepacked with Benzamidine Sepharose(R) 4 fast fLow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.