Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR CaptoTM Q Cytiva 1
Catalog No. 502904863
Encompass
HiTrap(R) Capto(TM) Q is a strong anion exchanger prepacked with BioProcess Capto(TM) media for screening and small-scale protein purifications using ion exchange chromatography (IEX).Capto(TM) Q has a strong quaternary ammonium (Q) anion exchanger coupled to a chemically modified high-flow agarose matrix. The modified agarose matrix provides greater particle rigidity without compromising pore size which allows more flexibility in process-scale applications. High dynamic binding capacity is achieved with a dextran surface extender that coats the agarose matrix.
Catalog No. 50-290-4863 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE11001303
HiTrap(R) Capto(TM) Q is a strong anion exchanger prepacked with BioProcess Capto(TM) media for screening and small-scale protein purifications using ion exchange chromatography (IEX).Capto(TM) Q has a strong quaternary ammonium (Q) anion exchanger coupled to a chemically modified high-flow agarose matrix. The modified agarose matrix provides greater particle rigidity without compromising pore size which allows more flexibility in process-scale applications. High dynamic binding capacity is achieved with a dextran surface extender that coats the agarose matrix.