Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Desalting Columns
Catalog No. 502904329
Encompass
HiTrap(R) Desalting is a ready-to-use 5 mL column prepacked with Sephadex(R) G-25 Superfine for fast and convenient desalting and buffer exchange.HiTrap(R) Desalting is a prepacked ready to use column for group separation between high and low molecular weight substances i.e. buffer exchange prior to or after different chromatographic steps removal of low molecular weight contaminants or removal of reagents to terminate a reaction.
Catalog No. 50-290-4329 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17140801
HiTrap(R) Desalting is a ready-to-use 5 mL column prepacked with Sephadex(R) G-25 Superfine for fast and convenient desalting and buffer exchange.HiTrap(R) Desalting is a prepacked ready to use column for group separation between high and low molecular weight substances i.e. buffer exchange prior to or after different chromatographic steps removal of low molecular weight contaminants or removal of reagents to terminate a reaction.