Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Hic Selection Kit
Catalog No. 502904237
Encompass
HiTrap(R) HIC Selection Kit consists of seven 1 mL columns prepacked with hydrophobic interaction chromatography media with different characteristics. Screen for the most appropriate HIC media and conditions to use for specific application and development work.
Catalog No. 50-290-4237 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28411007
HiTrap(R) HIC Selection Kit consists of seven 1 mL columns prepacked with hydrophobic interaction chromatography media with different characteristics. Screen for the most appropriate HIC media and conditions to use for specific application and development work.