Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR MabSelect SuReTM C
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE11003494
Encompass
HiTrap(R) MabSelect(TM) SuRe(TM) columns are prepacked with MabSelect(TM) SuRe a BioProcess medium for purification of monoclonal antibodies from large sample volumes. The novel alkali-tolerant recombinant Protein A ligand allow cleaning-in-place (CIP).HiTrap(R) MabSelect(TM) columns are prepacked with MabSelect(TM) a bioprocess medium for capuring of Mabs from large sample volumes.
Catalog No. 50-290-7284