Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR MabSelectTM Cytiva
Catalog No. 502909068
Encompass
HiTrap(R) MabSelect(TM) columns are prepacked with MabSelect a Bioprocess medium for capuring of Mabs from large sample volumes.
Catalog No. 50-290-9068 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28408256
HiTrap(R) MabSelect(TM) columns are prepacked with MabSelect a Bioprocess medium for capuring of Mabs from large sample volumes.